siRNA Details
| Gene Symbol: Ndrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011991.1 | NP_001011991.1 | 6, 7 | 523 | ||
| XM_006241708.2 | XP_006241770.1 | 6, 7 | 795 | |||
| GenBank mRNA | AY500369.1 | - | 6, 7 | 485 | ||
| BC081898.1 | - | 6, 7 | 523 | 7110455 | ||
| FQ212154.1 | - | 6, 7 | 535 | |||

Ndrg1
Genomic Map