siRNA Details
| Gene Symbol: Dscc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216934.8 | XP_216934.3 | 3 | 646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dscc1| Gene Symbol: Dscc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216934.8 | XP_216934.3 | 3 | 646 | ||