siRNA Details
| Gene Symbol: Mrpl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006985.1 | NP_001006986.1 | 5 | 366 | ||
| XM_006241651.1 | XP_006241713.1 | 5 | 561 | |||
| GenBank mRNA | BC084710.1 | - | 5 | 366 | 7302342 | |
| FQ220388.1 | - | 5 | 547 | |||
| FQ232270.1 | - | 5 | 410 | |||

Mrpl13
Genomic Map