siRNA Details
| Gene Symbol: Grhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134527.1 | NP_001127999.1 | 13, 14 | 1753 | ||
| XM_006241552.2 | XP_006241614.1 | 13, 14 | 2578 | |||
| GenBank mRNA | BC168688.1 | - | 13, 14 | 1753 | 7932577 | |

Grhl2
Genomic Map