siRNA Details
| Gene Symbol: Matn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216941.8 | XP_216941.7 | 1 | 169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Matn2| Gene Symbol: Matn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216941.8 | XP_216941.7 | 1 | 169 | ||