siRNA Details
| Gene Symbol: Lynx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130546.1 | NP_001124018.1 | 3 | 428 | ||
| GenBank mRNA | FQ212028.1 | - | 3 | 436 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lynx1| Gene Symbol: Lynx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130546.1 | NP_001124018.1 | 3 | 428 | ||
| GenBank mRNA | FQ212028.1 | - | 3 | 436 | ||