siRNA Details
| Gene Symbol: Rhpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241900.2 | XP_006241962.1 | 6, 7 | 745 | ||
| XM_006241901.2 | XP_006241963.1 | 6, 7 | 746 | |||
| XM_006241902.1 | XP_006241964.1 | 4, 5 | 359 | |||
| XM_008765628.1 | XP_008763850.1 | 6, 7 | 2134 | |||
| XM_216954.8 | XP_216954.4 | 6, 7 | 744 | |||

Rhpn1
Genomic Map