siRNA Details
| Gene Symbol: Zc3h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134865.1 | NP_001128337.1 | 4 | 1620 | ||
| XM_008765548.1 | XP_008763770.1 | 4 | 1642 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h3| Gene Symbol: Zc3h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134865.1 | NP_001128337.1 | 4 | 1620 | ||
| XM_008765548.1 | XP_008763770.1 | 4 | 1642 | |||