siRNA Details
| Gene Symbol: Tsta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127455.2 | NP_001120927.1 | 5 | 471 | ||
| NM_001270789.1 | NP_001257718.1 | 5 | 537 | |||
| NM_001270790.1 | NP_001257719.1 | 4 | 408 | |||
| GenBank mRNA | BC158885.1 | - | 4 | 380 | 8363456 | |
| BF289545.1 | - | 5 | 472 | |||
| CA339313.1 | - | 4 | 425 | |||
| CK653514.1 | - | 470 | ||||
| FQ210097.1 | - | 5 | 544 | |||
| FQ221078.1 | - | 5 | 459 | |||
| FQ229919.1 | - | 5 | 482 | |||

Tsta3
Genomic Map