siRNA Details
| Gene Symbol: Tsta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127455.2 | NP_001120927.1 | 3, 4 | 318 | ||
| NM_001270789.1 | NP_001257718.1 | 3, 4 | 384 | |||
| GenBank mRNA | BF289545.1 | - | 3, 4 | 319 | ||
| CA339313.1 | - | 2, 3 | 272 | |||
| CK653514.1 | - | 317 | ||||
| FQ210097.1 | - | 3, 4 | 391 | |||
| FQ221078.1 | - | 3, 4 | 306 | |||
| FQ229919.1 | - | 3, 4 | 329 | |||

Tsta3
Genomic Map