siRNA Details
| Gene Symbol: Tsta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127455.2 | NP_001120927.1 | 3 | 256 | ||
| NM_001270789.1 | NP_001257718.1 | 3 | 322 | |||
| NM_001270790.1 | NP_001257719.1 | 3 | 322 | |||
| GenBank mRNA | BC158885.1 | - | 3 | 294 | 8363456 | |
| BF289545.1 | - | 3 | 257 | |||
| CA339313.1 | - | 2 | 210 | |||
| CK653514.1 | - | 255 | ||||
| FQ210097.1 | - | 3 | 329 | |||
| FQ221078.1 | - | 3 | 244 | |||
| FQ229919.1 | - | 3 | 267 | |||

Tsta3
Genomic Map