siRNA Details
| Gene Symbol: Slc39a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077669.1 | NP_001071137.1 | 9 | 1566 | ||
| XM_008765563.1 | XP_008763785.1 | 10 | 3743 | |||
| XM_008765564.1 | XP_008763786.1 | 10 | 3621 | |||
| GenBank mRNA | BC127514.1 | - | 9 | 1566 | 7100017 | |

Slc39a4
Genomic Map