siRNA Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198752.3 | NP_942047.2 | 18 | 2286 | ||
| GenBank mRNA | AJ250329.1 | - | 18 | 2286 | ||
| AJ250330.1 | - | 17 | 2327 | |||
| FQ211833.1 | - | 16 | 2250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kifc2| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198752.3 | NP_942047.2 | 18 | 2286 | ||
| GenBank mRNA | AJ250329.1 | - | 18 | 2286 | ||
| AJ250330.1 | - | 17 | 2327 | |||
| FQ211833.1 | - | 16 | 2250 | |||