siRNA Details
| Gene Symbol: Ift27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130495.1 | NP_001123967.1 | 5 | 571 | ||
| XM_006241931.2 | XP_006241993.1 | 5 | 583 | |||
| GenBank mRNA | BC168150.1 | - | 581 | 7457249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift27| Gene Symbol: Ift27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130495.1 | NP_001123967.1 | 5 | 571 | ||
| XM_006241931.2 | XP_006241993.1 | 5 | 583 | |||
| GenBank mRNA | BC168150.1 | - | 581 | 7457249 | ||