siRNA Details
| Gene Symbol: Gga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011994.1 | NP_001011994.1 | 13 | 1573 | ||
| XM_006241973.2 | XP_006242035.1 | 14 | 2935 | |||
| XM_008765679.1 | XP_008763901.1 | 15 | 2691 | |||
| XM_008765680.1 | XP_008763902.1 | 12 | 2189 | |||
| XM_008765681.1 | XP_008763903.1 | 14 | 5397 | |||
| GenBank mRNA | BC090031.1 | - | 13 | 1573 | 7366311 | |

Gga1
Genomic Map