siRNA Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171981.1 | NP_001165452.1 | 3 | 143 | ||
| XM_008765682.1 | XP_008763904.1 | 2 | 1208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3bp1| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171981.1 | NP_001165452.1 | 3 | 143 | ||
| XM_008765682.1 | XP_008763904.1 | 2 | 1208 | |||