siRNA Details
| Gene Symbol: Fam227a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765683.1 | XP_008763905.1 | 7 | 1069 | ||
| GenBank mRNA | BC097470.1 | - | 7 | 800 | 7457750 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam227a| Gene Symbol: Fam227a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765683.1 | XP_008763905.1 | 7 | 1069 | ||
| GenBank mRNA | BC097470.1 | - | 7 | 800 | 7457750 | |