siRNA Details
Gene Symbol: Fam227a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765683.1 | XP_008763905.1 | 6, 7 | 989 | ||
GenBank mRNA | BC097470.1 | - | 6, 7 | 720 | 7457750 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam227a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765683.1 | XP_008763905.1 | 6, 7 | 989 | ||
GenBank mRNA | BC097470.1 | - | 6, 7 | 720 | 7457750 |