siRNA Details
| Gene Symbol: Dnal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009666.1 | NP_001009666.1 | 3 | 367 | ||
| GenBank mRNA | BC087688.1 | - | 3 | 367 | 7311257 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnal4| Gene Symbol: Dnal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009666.1 | NP_001009666.1 | 3 | 367 | ||
| GenBank mRNA | BC087688.1 | - | 3 | 367 | 7311257 | |