siRNA Details
| Gene Symbol: Slc25a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126269.1 | NP_001119741.1 | 6 | 590 | ||
| GenBank mRNA | BC166457.1 | - | 6 | 485 | 9025369 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a17| Gene Symbol: Slc25a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126269.1 | NP_001119741.1 | 6 | 590 | ||
| GenBank mRNA | BC166457.1 | - | 6 | 485 | 9025369 | |