siRNA Details
| Gene Symbol: Fam118a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173336.1 | NP_001166807.1 | 3 | 379 | ||
| XM_006242157.2 | XP_006242219.1 | 4 | 991 | |||
| XM_006242158.2 | XP_006242220.1 | 3 | 795 | |||
| XM_006242159.2 | XP_006242221.1 | 3 | 718 | |||
| XM_008765686.1 | XP_008763908.1 | 4 | 2138 | |||
| GenBank mRNA | BC169018.1 | - | 3 | 379 | 9028156 | |

Fam118a
Genomic Map