siRNA Details
Gene Symbol: Smc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130498.1 | NP_001123970.1 | 9 | 1457 | ||
XM_008765687.1 | XP_008763909.1 | 9 | 1492 |

More Information
Other Species Targeted

Gene Ontology Categories:
