siRNA Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134861.1 | NP_001128333.1 | 7 | 1003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cerk| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134861.1 | NP_001128333.1 | 7 | 1003 | ||