siRNA Details
| Gene Symbol: Trabd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106788.1 | NP_001100258.1 | 7 | 761 | ||
| XM_006242182.2 | XP_006242244.1 | 7 | 1323 | |||
| XM_006242183.2 | XP_006242245.1 | 7 | 1176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trabd| Gene Symbol: Trabd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106788.1 | NP_001100258.1 | 7 | 761 | ||
| XM_006242182.2 | XP_006242244.1 | 7 | 1323 | |||
| XM_006242183.2 | XP_006242245.1 | 7 | 1176 | |||