siRNA Details
| Gene Symbol: Ncaph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024877.1 | NP_001020048.1 | 12, 13 | 1106 | ||
| XM_006242186.2 | XP_006242248.1 | 11, 12 | 1068 | |||
| XM_006242187.2 | XP_006242249.1 | 11, 12 | 1068 | |||
| XM_006242188.2 | XP_006242250.1 | 11, 12 | 1029 | |||
| GenBank mRNA | BC097379.1 | - | 12, 13 | 1106 | 7445599 | |

Ncaph2
Genomic Map