siRNA Details
| Gene Symbol: Rpap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004243.1 | NP_001004243.1 | 6 | 721 | ||
| XM_008765690.1 | XP_008763912.1 | 7 | 1065 | |||
| GenBank mRNA | BC079414.1 | - | 6 | 721 | 7113336 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpap3| Gene Symbol: Rpap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004243.1 | NP_001004243.1 | 6 | 721 | ||
| XM_008765690.1 | XP_008763912.1 | 7 | 1065 | |||
| GenBank mRNA | BC079414.1 | - | 6 | 721 | 7113336 | |