siRNA Details
| Gene Symbol: Olr1102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765851.1 | XP_008764073.1 | 2 | 371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1102| Gene Symbol: Olr1102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765851.1 | XP_008764073.1 | 2 | 371 | ||