siRNA Details
| Gene Symbol: Olr1107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000424.1 | NP_001000424.1 | 1 | 229 | ||
| XM_006242320.2 | XP_006242382.1 | 2 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1107| Gene Symbol: Olr1107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000424.1 | NP_001000424.1 | 1 | 229 | ||
| XM_006242320.2 | XP_006242382.1 | 2 | 335 | |||