siRNA Details
| Gene Symbol: Kansl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004244.2 | NP_001004244.2 | 5 | 775 | ||
| XM_008765691.1 | XP_008763913.1 | 6 | 1103 | |||
| XM_008765692.1 | XP_008763914.1 | 6 | 859 | |||
| XM_008765693.1 | XP_008763915.1 | 6 | 859 | |||
| XM_008765694.1 | XP_008763916.1 | 4 | 675 | |||
| GenBank mRNA | BC079169.1 | - | 4 | 657 | 7129861 | |
| BC169120.1 | - | 5 | 1018 | |||
| EF076771.1 | - | 5 | 776 | |||
| Gene Symbol: LOC103690317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765853.1 | XP_008764075.1 | 1103 | |||
| XM_008765854.1 | XP_008764076.1 | 859 | ||||
| XM_008765855.1 | XP_008764077.1 | 859 | ||||
| XM_008765856.1 | XP_008764078.1 | 1063 | ||||
| XM_008765857.1 | XP_008764079.1 | 675 | ||||

Kansl2
Genomic Map