siRNA Details
| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013105.2 | NP_001013123.2 | 3 | 322 | ||
| XM_006257344.2 | XP_006257406.1 | 2 | 1227 | |||
| XM_008765695.1 | XP_008763917.1 | 2 | 225 | |||
| XM_008765696.1 | XP_008763918.1 | 2 | 237 | |||
| XM_008765697.1 | XP_008763919.1 | 2 | 214 | |||
| GenBank mRNA | BC084709.1 | - | 3 | 300 | 7301886 | |

Fkbp11
Genomic Map