siRNA Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191614.1 | NP_001178543.1 | 5 | 549 | ||
| XM_006257350.1 | XP_006257412.1 | 6 | 971 | |||
| XM_006257351.2 | XP_006257413.1 | 6 | 727 | |||
| XM_008765701.1 | XP_008763923.1 | 6 | 2617 | |||
| XM_008765702.1 | XP_008763924.1 | 6 | 1028 | |||

Spats2
Genomic Map