siRNA Details
| Gene Symbol: Smagp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182817.1 | NP_877969.1 | 3, 4 | 169 | ||
| XM_006242323.2 | XP_006242385.1 | 3, 4 | 298 | |||
| GenBank mRNA | AJ577837.1 | - | 3, 4 | 169 | ||
| BC101910.1 | - | 3, 4 | 484 | 7933976 | ||

Smagp
Genomic Map