siRNA Details
| Gene Symbol: Krt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047870.1 | NP_001041335.1 | 8 | 1204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt7| Gene Symbol: Krt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047870.1 | NP_001041335.1 | 8 | 1204 | ||