siRNA Details
| Gene Symbol: Krt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008802.2 | NP_001008802.2 | 4 | 932 | ||
| GenBank mRNA | BC127464.1 | - | 4 | 932 | 8370075 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt1| Gene Symbol: Krt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008802.2 | NP_001008802.2 | 4 | 932 | ||
| GenBank mRNA | BC127464.1 | - | 4 | 932 | 8370075 | |