siRNA Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106795.1 | NP_001100265.1 | 6 | 668 | ||
| XM_006242387.2 | XP_006242449.1 | 5 | 585 | |||
| XM_008765704.1 | XP_008763926.1 | 5 | 628 | |||
| XM_008765705.1 | XP_008763927.1 | 5 | 609 | |||

Aaas
Genomic Map