siRNA Details
| Gene Symbol: ZNF214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013249.2 | NP_037381.2 | 3 | 2063 | ||
| XM_005253128.3 | XP_005253185.1 | 4 | 2138 | |||
| XM_005253129.3 | XP_005253186.1 | 3 | 2071 | |||
| XM_006718308.3 | XP_006718371.1 | 3 | 2071 | |||
| XM_006718309.3 | XP_006718372.1 | 3 | 2067 | |||
| XM_011520355.2 | XP_011518657.1 | 4 | 2134 | |||
| XM_011520357.2 | XP_011518659.1 | 3 | 1825 | |||
| XM_017018269.1 | XP_016873758.1 | 3 | 1839 | |||
| GenBank mRNA | AF056617.1 | - | 3 | 2018 | ||
| AK313462.1 | - | 3 | 2035 | |||
| BC136772.1 | - | 3 | 1943 | 9020762 | ||
| BC144405.1 | - | 4 | 2002 | 9052926 | ||

ZNF214
Genomic Map