siRNA Details
| Gene Symbol: AQP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001651.3 | NP_001642.1 | 3 | 1108 | ||
| XM_005268838.2 | XP_005268895.1 | 3 | 749 | |||
| GenBank mRNA | BC032946.1 | - | 3 | 658 | 5269384 | |
| CR541989.1 | - | 3 | 586 | |||
| CR542022.1 | - | 3 | 586 | |||

AQP5
Genomic Map