siRNA Details
Gene Symbol: LOC300308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013952.1 | NP_001013974.1 | 834 | |||
GenBank mRNA | BC090074.1 | - | 834 | 7114534 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC300308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013952.1 | NP_001013974.1 | 834 | |||
GenBank mRNA | BC090074.1 | - | 834 | 7114534 |