siRNA Details
| Gene Symbol: L3mbtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033695.1 | NP_001028867.1 | 8 | 952 | ||
| XM_008765711.1 | XP_008763933.1 | 8 | 1032 | |||
| GenBank mRNA | BC101865.1 | - | 8 | 952 | 7938856 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

L3mbtl2| Gene Symbol: L3mbtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033695.1 | NP_001028867.1 | 8 | 952 | ||
| XM_008765711.1 | XP_008763933.1 | 8 | 1032 | |||
| GenBank mRNA | BC101865.1 | - | 8 | 952 | 7938856 | |