siRNA Details
| Gene Symbol: LOC100910997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242486.2 | XP_006242548.2 | 732 | |||
| Gene Symbol: Mmp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765897.1 | XP_008764119.1 | 5 | 732 | ||

LOC100910997
Genomic Map