siRNA Details
| Gene Symbol: Cwc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024987.1 | NP_001020158.1 | 6 | 600 | ||
| XM_006242520.2 | XP_006242582.1 | 6 | 631 | |||
| GenBank mRNA | BC091396.1 | - | 6 | 600 | 7323256 | |
| FQ224752.1 | - | 6 | 570 | |||

Cwc15
Genomic Map