siRNA Details
| Gene Symbol: Med17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106801.1 | NP_001100271.1 | 8 | 1365 | ||
| GenBank mRNA | BC083809.1 | - | 7 | 1343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med17| Gene Symbol: Med17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106801.1 | NP_001100271.1 | 8 | 1365 | ||
| GenBank mRNA | BC083809.1 | - | 7 | 1343 | ||