siRNA Details
| Gene Symbol: Olr1146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766032.1 | XP_008764254.1 | 3 | 1129 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1146| Gene Symbol: Olr1146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766032.1 | XP_008764254.1 | 3 | 1129 | ||