siRNA Details
| Gene Symbol: Olr1155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000433.1 | NP_001000433.1 | 1 | 710 | ||
| Gene Symbol: Olr1149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000874.1 | NP_001000874.1 | 1 | 743 | ||

Olr1155
Genomic Map