siRNA Details
| Gene Symbol: Olr1169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000434.1 | NP_001000434.1 | 1 | 592 | ||
| Gene Symbol: LOC100911547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750445.3 | XP_003750493.2 | 604 | |||

Olr1169
Genomic Map