siRNA Details
Gene Symbol: ZNF223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013361.5 | NP_037493.3 | 5 | 1398 | ||
XM_017027258.1 | XP_016882747.1 | 5 | 1643 | |||
XM_017027259.1 | XP_016882748.1 | 5 | 1698 | |||
XM_017027260.1 | XP_016882749.1 | 2 | 2617 | |||
GenBank mRNA | AK292327.1 | - | 5 | 1361 | ||
BC022466.1 | - | 5 | 1398 | 4793666 |
Gene Symbol: ZNF284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308295.1 | - | 7 | 1558 |
