siRNA Details
| Gene Symbol: Lppr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005881.3 | NP_001005881.2 | 6 | 1025 | ||
| XM_006242628.2 | XP_006242690.1 | 6 | 1394 | |||
| XM_006242629.2 | XP_006242691.1 | 4 | 531 | |||
| XM_008765930.1 | XP_008764152.1 | 6 | 1319 | |||
| XM_008765931.1 | XP_008764153.1 | 5 | 1205 | |||

Lppr2
Genomic Map