siRNA Details
| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006986.1 | NP_001006987.1 | 3 | 450 | ||
| XM_006242636.2 | XP_006242698.1 | 3 | 497 | |||
| XM_006242637.2 | XP_006242699.1 | 3 | 486 | |||
| XM_006242638.2 | XP_006242700.1 | 3 | 485 | |||
| GenBank mRNA | BC083762.1 | - | 3 | 450 | 7193076 | |
| FQ209658.1 | - | 3 | 458 | |||
| FQ212584.1 | - | 3 | 481 | |||
| FQ215401.1 | - | 3 | 461 | |||

Ecsit
Genomic Map