siRNA Details
| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006986.1 | NP_001006987.1 | 7 | 1092 | ||
| XM_006242636.2 | XP_006242698.1 | 7 | 1157 | |||
| XM_006242637.2 | XP_006242699.1 | 7 | 1146 | |||
| XM_006242638.2 | XP_006242700.1 | 7 | 1127 | |||
| GenBank mRNA | BC083762.1 | - | 7 | 1092 | 7193076 | |
| FQ209658.1 | - | 7 | 1100 | |||
| FQ212584.1 | - | 7 | 1123 | |||
| FQ215401.1 | - | 7 | 1121 | |||

Ecsit
Genomic Map