siRNA Details
| Gene Symbol: Npsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106808.1 | NP_001100278.1 | 7 | 785 | ||
| XM_008766049.1 | XP_008764271.1 | 6 | 1028 | |||
More Information
Other Species Targeted
Set Membership: |
|

Npsr1
Genomic Map